missing translation for 'onlineSavingsMsg'
Learn More

Robu™ Micro-Filter-Candles

Robu™ Micro-Filter-Candles are highly resistant to most chemicals such as solvents, acids, and alkalis.

Brand:  Robu™ 18123

Additional Details : Weight : 0.11540kg

 View more versions of this product

Product Code. 16822946

  • 2285.00 SEK / Pack of 5
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

  • ISO 4793-80
  • Cylindrical shape
  • With narrow glass tube
Specifications

Specifications

Cylindrical
Narrow glass tube
210 mm
13 mm
16 to 40 μm
Filter Candle
25 mm
8 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers