Learn More
Ansell MICROFLEX™ 93-864 MidKnight™ MAX Disposable Nitrile Gloves - Extended Cuff
Heavy duty gloves mask dirt and stains, in addition to providing expanded protection with an extended cuff. Additional thickness in the palm helps to prevent tearing during use, while textured surfaces enhances grip capabilities.
244.00 SEK - 390.00 SEK
Specifications
Certifications/Compliance (Europe) | EN 420:2003 + A1:2009 |
---|---|
AQL | 1.5 |
Color | Black |
Disposable | Single-use |
Finger Thickness (Metric) | 0.21 mm |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
16609123
|
Ansell
93864070 |
7 | N/A | |||||||
16619123
|
Ansell
93864080 |
8 |
390.00 SEK
Pack of 100 |
In Stock
Log in to see stock. |
||||||
16629123
|
Ansell
93864090 |
9 | N/A | |||||||
16639123
|
Ansell
93864100 |
10 | N/A | |||||||
16649123
|
Ansell
93864110 |
11 | N/A | |||||||
Description
- Additional palm thickness (0.16 mm) resists tearing during use
- Deep black color hides the appearance of oils, grease, and grime
- Extended cuff provides extra protection over the wrist and forearm
- Resistant to a range of chemicals, oils, and solvents
- Fully textured design enhances grip capabilities
- Silicone free design is paint and finish process friendly
Specifications
EN 420:2003 + A1:2009 | |
Black | |
0.21 mm | |
Nitrile | |
Powder Free | |
JKPT | |
39-864 |
1.5 | |
Single-use | |
Ambidextrous | |
0.16 mm | |
Type B | |
Beaded | |
300 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Ansell MICROFLEX™ 93-864 MidKnight™ MAX Disposable Nitrile Gloves - Extended Cuff