Learn More
Kern Sohn Microscope Camera ODC
Specialists in microscopy for measurement, counting, documentation, archiving and image processing.
4814.94 SEK - 27128.05 SEK
Product Code | Brand | Port Type | Resolution | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Port Type | Resolution | Size | Price | Quantity & Availability | ||||
17309541
![]() |
Kern Sohn
ODC 825 |
USB 2.0 | 5.1 MP | 1/2.5 in. Sensor |
4814.94 SEK
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||
17319541
![]() |
Kern Sohn
ODC 832 |
USB 3.0 | 5.1 MP | 1/2.5 in. Sensor |
6576.50 SEK
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||
17329541
![]() |
Kern Sohn
ODC 841 |
USB 2.0 | 20 MP | 1 in. Sensor |
14914.56 SEK
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||
17339541
![]() |
Kern Sohn
ODC 861 |
USB 2.0 | 20 MP | Compact, 1 in. Sensor |
27128.05 SEK
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||
Description
- The universal microscope cameras can be used anywhere and can be connected to the microscope as well as to a laptop or PC using the USB cable (USB 2.0)
- Through the proven CMOS technology, in connection with the USB 2.0 the images are shown quickly and clearly
- These cameras are also ideal for more demanding applications, such as, for example, darkfield, phase contrast and for fluorescence applications.
- Your daily work is made significantly easier with the very best synchronisation, a high frame rate as well as stable image performance together with our Microscope VIS software which we deliver with the product.
- The power supply is through the USB cable, which means that no additional power supply is required
- As well as the camera, the delivery includes our multi-lingual KERN Microscope VIS Basic KERN OXM 901, a USB cable (Length: 2000 mm) various eyepiece adapters and an object micrometer to calibrate the software.
- Please order the appropriate C-mount adapter to fit your KERN microscope now.
Specifications
Microscopes | |
CMOS |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.