missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Midkine Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | Midkine |
---|---|
Concentration | 233 ÎĽg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
CĂłdigo de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
CĂłdigo de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
16886185
|
Proteintech
28546-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16876185
|
Proteintech
28546-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
DescripciĂłn
The Midkine antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Midkine. This antibody recognizes human, mouse antigen. The Midkine antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
Midkine | |
Western Blot, ELISA | |
Unconjugated | |
Human, Mouse | |
IgG |
233 ÎĽg/mL | |
Polyclonal | |
Rabbit | |
4192 | |
Primary |
ÂżDetecta una oportunidad de mejora?Comparta una correcciĂłn de contenido
CorrecciĂłn del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
TĂtulo del producto
Midkine Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech