Learn More
Cytiva Mini Profile™ Capsules with Profile Star Filter
Mini Profile™ Capsules designed for research applications only.
Brand: Cytiva 12071
This item is not returnable.
View return policy
Description
- Users have the convenient choice between high void-volume depths medium or high-area pleated depth filter
- Modules can come in different filter specifications to meet the specific needs of an early phase process development
Specifications
Spun Polypropylene | |
Non-sterile | |
3/Pk. |
3 μm | |
90 cm2 | |
9/16 in. Hose Barb |
Product Content Correction
Product Title
Cytiva Mini Profile™ Capsules with Profile Star Filter > 3 μm
Spot an opportunity for improvement?Share a Content Correction