Learn More
Trajan Scientific and Medical™ Mini Union Replacement Ferrules
Delivers an easy, leak-free installation for capillary columns without the use of any tools.
Brand:  Trajan Scientific and Medical™ 072694
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15248903
  • 1155.00 SEK / Pack of 10
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
Description
Description
  • Specifically designed for connecting capillary columns
  • Connects guard columns to capillary columns
  • Inert and precise glass lined bore
  • Low dead volume
Specifications
Specifications
Bore size: No Hole
15% Graphite, 85% Vespel
Graphite Vespel Replacement Mini Union Ferrule No-Hole
10/Pk.