Learn More
Trajan Scientific and Medical™ Mini Union Replacement Ferrules
Delivers an easy, leak-free installation for capillary columns without the use of any tools.
1155.00 SEK
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
13457349
|
Trajan Scientific and Medical™
072697 |
Bore size: 0.5 mm |
1155.00 SEK
Pack of 10 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
15268903
|
Trajan Scientific and Medical™
072698 |
Bore size: 0.8 mm |
1155.00 SEK
Pack of 10 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
15258903
|
Trajan Scientific and Medical™
072695 |
Bore size: 0.3 mm |
1155.00 SEK
Pack of 10 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
15248903
|
Trajan Scientific and Medical™
072694 |
Bore size: No Hole |
1155.00 SEK
Pack of 10 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
Description
- Specifically designed for connecting capillary columns
- Connects guard columns to capillary columns
- Inert and precise glass lined bore
- Low dead volume
Specifications
15% Graphite, 85% Vespel | |
10/Pk. |