Läs mer
Minoxidil, Thermo Scientific™
Minoxidil, CAS # 38304-91-5, is a comopund with vasodilating effects that works by opening adenosine triphosphate (ATP)-sensitive potassium channels.
Brand:  Thermo Scientific Alfa Aesar J61803.03
Kvantitet 1g
 Se fler versioner av denna produkt
Produktkod. 15444319
  • 452.91 SEK / 1g
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Denne vare kan ikke returneres. Se returpolicy
Beskrivelse
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Minoxidil is a piperidine that acts as a vasodilator when converted into its active metabolite, minoxidil sulphate
• Minoxidil opens adenosine triphosphate (ATP)-sensitive potassium channels in the plasma membrane and relaxes smooth muscle in arteriole walls. Relaxation of smooth muscle within the arterioles leads to decreased resistance and lower blood pressure
Application
• Minoxidil metabolism is a common focus of its use in research because of the number of enzymes involved in its sulfation and clearance
• Minoxidil can be used for in vivo or in vitro physiology research
TRUSTED_SUSTAINABILITY
Tekniske data
Kemiske identifikatorer
38304-91-5
209.253
ZIMGGGWCDYVHOY-UHFFFAOYSA-N
4201
C1CCN(CC1)C2=NC(=N)N(C(=C2)N)O
C9H15N5O
MFCD00063409
minoxidil, rogaine, loniten, regaine, minoximen, theroxidil, tricoxidil, alopexil, alostil, lonolox
3-hydroxy-2-imino-6-piperidin-1-ylpyrimidin-4-amine
Tekniske data
Minoxidil
C9H15N5O
1g
minoxidil, rogaine, loniten, regaine, minoximen, theroxidil, tricoxidil, alopexil, alostil, lonolox
ZIMGGGWCDYVHOY-UHFFFAOYSA-N
3-hydroxy-2-imino-6-piperidin-1-ylpyrimidin-4-amine
4201
38304-91-5
MFCD00063409
14,6203
Soluble in water (2.2mg/ml),100% ethanol (29mg/ml),propylene glycol,acetone,DMSO (6.5mg/ml),and methanol.
C1CCN(CC1)C2=NC(=N)N(C(=C2)N)O
209.253
209.25
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Produktcertifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.