missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIP-3-beta Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13397-1-AP-20UL
This item is not returnable.
View return policy
Description
Macrophage Inflammatory Protein-3 beta (MIP-3 beta), also called CCL19, is a chemokine that is expressed in the thymus, lymph nodes and activated bone marrow stromal cells. MIP-3 beta signals through the G protein-coupled receptor, CCR7, to regulate normal lymphocyte recirculation and homing of T and B cells to the lymph nodes. Human MIP-3 beta shows activity on mouse cells.Specifications
MIP-3-beta | |
Polyclonal | |
Unconjugated | |
CCL19 | |
Beta chemokine exodus 3, C C motif chemokine 19, CCL19, CK beta 11, CKb11, EBI1 ligand chemokine, ELC, MIP 3 beta, MIP 3b, MIP3B, MIP-3-beta, SCYA19, Small inducible cytokine A19 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6363 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99731 | |
CCL19 | |
MIP-3-beta Fusion Protein Ag4200 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MIP-3-beta Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction