Learn More
MIS12 Mouse anti-Human, Unconjugated, Clone: 2G5F6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67475-1-IG-150UL
 View more versions of this product
Product Code. 16890116
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The MIS12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MIS12. This antibody recognizes human antigen. The MIS12 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
MIS12
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
2G5F6
Unconjugated
150 μL
79003
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MIS12 Mouse anti-Human, Unconjugated, Clone: 2G5F6, Proteintech > 150 μL