missing translation for 'onlineSavingsMsg'
Learn More
ML-7 hydrochloride, MedChemExpress Product Code.: 16482597

ML-7 hydrochloride, MedChemExpress

Product Code. 16482597
50 mg, 50mg
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16482597

Brand: Medchem Express HY15417

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ML-7 hydrochloride is a naphthalene sulphonamide derivative, potently inhibits MLCK (IC50=300 nM). ML-7 hydrochloride also inhibits YAP/TAZ.

ML-7 hydrochloride is a naphthalene sulphonamide derivative, potently inhibits MLCK (IC50=300 nM). ML-7 hydrochloride also inhibits YAP/TAZ.

Specifications

Chemical Name or Material ML-7 hydrochloride
CAS 110448-33-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H18ClIN2O2S
Quantity 50 mg
Solubility Information DMSO : ≥ 43 mg/mL (94.98 mM) ∣H2O : 1.43 mg/mL (3.16 mM; Need ultrasonic)
SMILES O=S(N1CCNCCC1)(C2=C3C=CC=C(I)C3=CC=C2)=O.[H]Cl
Molecular Weight (g/mol) 452.74
Formula Weight 452.74
Percent Purity 99.49%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
ML-7 hydrochloride, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.