missing translation for 'onlineSavingsMsg'
Learn More

MLKL Mouse anti-Human, Mouse, Clone: 3D4C6, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66675-1-IG-20UL

 View more versions of this product

Product Code. 16868775

  • 1645.54 SEK / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.
Specifications

Specifications

MLKL
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8NB16, Q9D2Y4
Mlkl
MLKL Fusion Protein Ag15188
20 μL
Primary
Mouse, Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
3D4C6
Unconjugated
MLKL
FLJ34389, MLKL
Mouse
Protein G
RUO
197259, 74568
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MLKL Mouse anti-Human, Mouse, Clone: 3D4C6, Proteintech > 20 μL; Unconjugated

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.