missing translation for 'onlineSavingsMsg'
Learn More

MLKL Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen MLKL
Concentration 0.35 mg/mL
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16890665
View Documents
Proteintech
21066-1-AP-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16880665
View Documents
Proteintech
21066-1-AP-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.
Specifications

Specifications

MLKL
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human
Q8NB16
197259
MLKL Fusion Protein Ag15188
Primary
-20°C
MLKL
0.35 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
BC028141, MLKL
MLKL
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MLKL Rabbit anti-Human, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.