missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP21 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55289-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to MMP21.
This gene encodes a member of the matrix metalloproteinase family. Proteins in this family are involved in the breakdown of extracellular matrix for both normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, and disease processes, such as asthma and metastasis. The encoded protein may play an important role in embryogenesis, particularly in neuronal cells, as well as in lymphocyte development and survival.Specifications
MMP21 | |
Polyclonal | |
Unconjugated | |
MMP21 | |
matrix metallopeptidase 21, Matrix metalloproteinase 21, MMP 21, MMP21 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
118856, 214766 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K3F2, Q8N119 | |
Mmp21 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MMP21 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction