Learn More
MMP3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17873-1-AP-150UL
 View more versions of this product
Product Code. 16887864
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The MMP3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MMP3. This antibody recognizes human, mouse, rat antigen. The MMP3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
MMP3
Polyclonal
Unconjugated
150 μL
4314
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA
200 μg/mL
Rabbit
Primary
Human, Rat, Mouse
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MMP3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 150 μL