missing translation for 'onlineSavingsMsg'
Learn More

MNT Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1355.00 SEK - 3665.00 SEK

Specifications

Antigen MNT
Concentration 0.31 mg/mL
Applications Immunocytochemistry, Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16872425
View Documents Promotion Details
Proteintech
23742-1-AP-150UL
150 μL
3665.00 SEK
150µL
View alternative products
Save Up to 
»
Estimated Shipment: 12-06-2023
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16882425
View Documents Promotion Details
Proteintech
23742-1-AP-20UL
20 μL
1355.00 SEK
20µL
View alternative products
Save Up to 
»
Estimated Shipment: 12-06-2023
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Myc/Max/Mad network comprises a group of transcription factors that co-interact to regulate gene-specific transcriptional activation or repression. This gene encodes a protein member of the Myc/Max/Mad network. This protein has a basic-Helix-Loop-Helix-zipper domain (bHLHzip) with which it binds the canonical DNA sequence CANNTG, known as the E box, following heterodimerization with Max proteins. This protein is likely a transcriptional repressor and an antagonist of Myc-dependent transcriptional activation and cell growth. This protein represses transcription by binding to DNA binding proteins at its N-terminal Sin3-interaction domain.
Specifications

Specifications

MNT
Immunocytochemistry, Western Blot, Immunofluorescence
Unconjugated
Rabbit
Human
Q99583
4335
MNT Fusion Protein Ag20666
Primary
-20°C
MNT
0.31 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
bHLHd3, MAD6, MAX binding protein, Max binding protein MNT, MNT, MXD6, Myc antagonist MNT, Protein ROX, ROX
MNT
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications