missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lectin SBA From Glycine max (soybean), Alexa Fluor™ 647 Conjugate

Suitable for use in cell tracing and tracking, cellular imaging and flow cytometry

Brand:  Invitrogen™ L32463

Product Code. 10563093

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

LOLO™-1 iodide (565/579). 1mM solution in DMSO.

Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, Flow Cytometry, Flow Cytometry Antibodies & Secondary Detection, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining & Detection, In Vivo Imaging, Microbial Tracking

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 mg
Alexa Fluor 647
Alexa Fluor Dyes
Lyophilized
Alexa Fluor™
Soybean
Store in freezer (-5 to -30°C) and protect from light.
Room Temperature
Lectins
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2529346 Certificate TypeCertificate of Analysis Date12/09/2022 Catalog NumberL32463
Lot Number2411976 Certificate TypeCertificate of Analysis Date13/10/2021 Catalog NumberL32463
Lot Number2265141 Certificate TypeCertificate of Analysis Date29/09/2020 Catalog NumberL32463
Lot Number1891287 Certificate TypeCertificate of Analysis Date19/07/2017 Catalog NumberL32463
Lot Number1904163 Certificate TypeCertificate of Analysis Date19/07/2017 Catalog NumberL32463
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Lectin SBA From Glycine max (soybean), Alexa Fluor™ 647 Conjugate > 1mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.