Learn More
Merck Montage™ SEQ96 Sequencing Reaction Cleanup Kit
Use 96-well filter plates with proprietary size-exclusion membranes to retain sequencing products while excess dye terminator is removed quickly and efficiently. Fully automatable kits rinse away unincorporated dye terminator without the need for binding and eluting protocols.
Brand:  Merck LSKS09624
 View more versions of this product
Product Code. 12390180
  • 41350.00 SEK / Pack of 24
Includes: Clear polystyrene 96-well plates, optimized injection solution
Requires : MultiScreen™ 384 Vacuum Manifold with underdrain support grid (Mfr. No. MAVM 096 0R)
Description
Description
Use 96-well filter plates with proprietary size-exclusion membranes to retain sequencing products while excess dye terminator is removed quickly and efficiently. Fully automatable kits rinse away unincorporated dye terminator without the need for binding and eluting protocols.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DNA sequencing
96-well filter plates and Injection solution
24 Pack
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Merck Montage™ SEQ96 Sequencing Reaction Cleanup Kit > DNA Purification Kit; 24/Pk.