Learn More
Merck Montage™ SEQ96 Sequencing Reaction Cleanup Kit
Use 96-well filter plates with proprietary size-exclusion membranes to retain sequencing products while excess dye terminator is removed quickly and efficiently. Fully automatable kits rinse away unincorporated dye terminator without the need for binding and eluting protocols.
Brand: Merck LSKS09624
Includes: Clear polystyrene 96-well plates, optimized injection solution
Requires : MultiScreen™ 384 Vacuum Manifold with underdrain support grid (Mfr. No. MAVM 096 0R)
This item is not returnable. View return policy
Description
Use 96-well filter plates with proprietary size-exclusion membranes to retain sequencing products while excess dye terminator is removed quickly and efficiently. Fully automatable kits rinse away unincorporated dye terminator without the need for binding and eluting protocols.
Specifications
DNA sequencing | |
96-well filter plates and Injection solution | |
24 Pack |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.