missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitellogenin (VTG), Mouse, Clone: 5A4, Abnova™
Mouse monoclonal antibody raised against native vitellogenin.
Brand: Abnova MAB1730.100ug
This item is not returnable.
View return policy
Specifications
Vitellogenin (VTG) | |
Monoclonal | |
Unconjugated | |
Western Blot (0.1-1 ug/mL) ELISA (0.25-2 ug/mL) The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μg | |
Primary | |
Liquid |
ELISA, Western Blot | |
5A4 | |
Mouse monoclonal antibody raised against native vitellogenin. | |
In PBS (0.1% BSA, 0.1% proclin) | |
Native purified Red Sea Bream vitellogenin. | |
RUO | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vitellogenin (VTG), Mouse, Clone: 5A4, Abnova™ > 100μg; Unlabeled
Spot an opportunity for improvement?Share a Content Correction