Learn More
Novus Biologicals™ Mouse Gremlin 1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand:  Novus Biologicals™ NBP2-75366
Product Code. 18337220
  • 8125.00 SEK / 1 set
Description
The Mouse Gremlin 1 ELISA Kit (Colorimetric) measures mouse Gremlin 1 in serum, plasma and other biological fluids.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
78.13 - 5000 pg/mL
This kit recognizes Mouse GREM1 in samples. No significant cross-reactivity or interference between Mouse GREM1 and analogues was observed.
Biological Fluids, Plasma, Serum
26585
Mouse
Storage of components varies. See protocol for specific instructions.
46.88 pg/mL
ELISA Kit (Colorimetric)
ELISA
GREM1
1 Kit
Gremlin 1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Novus Biologicals™ Mouse Gremlin 1 ELISA Kit (Colorimetric) > Quantity: 1 Kit; Host Species: Mouse