Learn More
Novus Biologicals™ Mouse Gremlin 1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand: Novus Biologicals™ NBP2-75366
This item is not returnable.
View return policy
Description
The Mouse Gremlin 1 ELISA Kit (Colorimetric) measures mouse Gremlin 1 in serum, plasma and other biological fluids.
Specifications
78.13 - 5000 pg/mL | |
This kit recognizes Mouse GREM1 in samples. No significant cross-reactivity or interference between Mouse GREM1 and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
26585 | |
Mouse | |
Storage of components varies. See protocol for specific instructions. |
46.88 pg/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
GREM1 | |
1 Kit | |
Gremlin 1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Mouse Gremlin 1 ELISA Kit (Colorimetric) > Quantity: 1 Kit; Host Species: Mouse
Spot an opportunity for improvement?Share a Content Correction