Learn More
Novus Biologicals™ Mouse Haptoglobin ELISA Kit (Colorimetric)
Brand:  Novus Biologicals™ KA1921
Product Code. 18613420
  • 11245.00 SEK / 1 set
Specifications
1.95 to 125 ng/mL
ELISA Kit (Colorimetric)
ELISA, Quantification
HP
Cardiovascular Biology
binding peptide, BP, haptoglobin, haptoglobin alpha(1 S)-beta, haptoglobin alpha(2 FS)-beta, haptoglobin, alpha polypeptide, haptoglobin, beta polypeptide, HP2ALPHA2, HPA1S, MGC111141, zonulin
0.551
Plasma, Serum
3240
1 Kit
Store at 4°C.
Mouse
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Novus Biologicals™ Mouse Haptoglobin ELISA Kit (Colorimetric) > 1 Kit