Learn More
Novus Biologicals™ Mouse Haptoglobin ELISA Kit (Colorimetric)
Brand: Novus Biologicals™ KA1921
This item is not returnable.
View return policy
Description
Specifications
1.95 to 125 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA, Quantification | |
HP | |
Cardiovascular Biology | |
binding peptide, BP, haptoglobin, haptoglobin alpha(1 S)-beta, haptoglobin alpha(2 FS)-beta, haptoglobin, alpha polypeptide, haptoglobin, beta polypeptide, HP2ALPHA2, HPA1S, MGC111141, zonulin |
0.551 | |
Plasma, Serum | |
3240 | |
1 Kit | |
Store at 4°C. | |
Mouse |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Mouse Haptoglobin ELISA Kit (Colorimetric) > 1 Kit
Spot an opportunity for improvement?Share a Content Correction