missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IFN-beta Antibody, R&D Systems™

Rat Monoclonal Antibody
1875.00 SEK - 4405.00 SEK
Specifications
Antigen | IFN-beta |
---|---|
Clone | 944233 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18761817
![]() |
R&D Systems
MAB82342-SP |
25 ÎĽg |
1875.00 SEK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18732447
![]() |
R&D Systems
MAB82342-100 |
100 ÎĽg |
4405.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-beta Monoclonal antibody specifically detects IFN-beta in Mouse samples. It is validated for ELISASpecifications
IFN-beta | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 ÎĽm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 ÎĽm filtered solution in PBS. | |
Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 | |
Human embryonic kidney cell HEK293-derived Mouse IFN-beta , Ile22-Asn182, Accession # P01575 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
944233 | |
ELISA | |
Unconjugated | |
Rat | |
Mouse | |
P01575 | |
3456 | |
IgG2b | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mouse IFN-beta Antibody, R&D Systems™