missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Leptin/OB Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems BAF498
This item is not returnable.
View return policy
Description
Leptin/OB Polyclonal specifically detects Leptin/OB in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
Leptin/OB | |
Polyclonal | |
Biotin | |
Q544U0 | |
LEP | |
E. coli-derived recombinant mouse Leptin (R&D Systems, Catalog # 498-OB) Val22-Cys167 Accession # Q544U0 | |
50 μg | |
Primary | |
Detects mouse Leptin/OB in ELISAs and Western blots. In sandwich immunoassays, less than 45% cross-reactivity with recombinant rat (rr) Leptin is observed and less than 0.2% cross-reactivity with rhLeptin and rmLeptin R is observed. | |
Mouse | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
FLJ94114, LEP, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), OB, Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS | |
Goat | |
Affinity Purified | |
RUO | |
3952 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mouse Leptin/OB Biotinylated Antibody, R&D Systems™ > 50μg; Biotin
Spot an opportunity for improvement?Share a Content Correction