missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Mouse Nectin-4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3116-SP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Nectin-4/PVRL4 Polyclonal specifically detects Nectin-4/PVRL4 in Mouse samples. It is validated for Western Blot.
Specifikationer
| Nectin-4/PVRL4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EDSS1, Ig superfamily receptor LNIR, LNIR, LNIRnectin 4, nectin-4, Nectin4, poliovirus receptor-related 4, poliovirus receptor-related protein 4, PRR4, PRR4DKFZp686K05193, PVRL4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 81607 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q8R007 | |
| PVRL4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Nectin-4 Tyr28-Ile349 Accession # Q8R007 | |
| 25 μg | |
| Primary | |
| Detects mouse Nectin-4 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human Nectin-4 is observed and less than 5% cross-reactivity with recombinant mouse Nectin-1, -2, and -3 is observed. | |
| Mouse | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering