missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Osteopontin/OPN PE-conjugated Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems IC808P
This item is not returnable.
View return policy
Description
Osteopontin/OPN Polyclonal specifically detects Osteopontin/OPN in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
Osteopontin/OPN | |
Polyclonal | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
BNSP, Bone sialoprotein 1, Eta-1, MGC110940, Nephropontin, OPN, osteopontin, secreted phosphoprotein 1bone sialoprotein I, early T-lymphocyteactivation 1), secreted phosphoprotein-1 (osteopontin, bone sialoprotein), Spp1, SPP-1, SPP1/CALPHA1 fusion, Urinary stone protein, uropontin | |
Goat | |
Affinity Purified | |
RUO | |
6696 | |
Mouse | |
IgG |
Flow Cytometry | |
R-PE | |
Q547B5 | |
SPP1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Osteopontin/OPN Leu17-Asn294 (Glu99Gly) Accession # Q547B5 | |
100 Tests | |
Primary | |
Detects mouse Osteopontin (OPN) in ELISAs and Western blots. In sandwich ELISAs, less than 3% cross-reactivity with recombinant human OPN is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mouse Osteopontin/OPN PE-conjugated Antibody, R&D Systems™ > 100 Tests; PE
Spot an opportunity for improvement?Share a Content Correction