missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Mouse PD-L1/B7-H1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB90783-100
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
PD-L1 Monoclonal specifically detects PD-L1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.
Specifikationer
| PD-L1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse PD-L1/B7-H1, Phe19-His239, Accession No. Q9EP73 | |
| 100 μg | |
| 29126.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Flow Cytometry, Blocking Assay, CyTOF | |
| 2341A | |
| Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
| Q9EP73 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects mouse PD-L1/B7-H1 in direct ELISAs. | |
| Mouse | |
| Supernatant |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering