missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Mouse/Rabbit IgG VisUCyte HRP Polymer Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems VC002-025
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
IgG Polyclonal specifically detects IgG in Mouse, Rabbit samples. It is validated for Immunohistochemistry.
Specifikationer
| IgG | |
| Polyclonal | |
| Immunohistochemistry | |
| Immunoglobulin G, ImmunoglobulinG | |
| Mouse and Rabbit IgG | |
| 25 mL | |
| Secondary | |
| Detects heavy ( gamma ) chains on mouse and rabbit IgG and light chains on all mouse and rabbit immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin mouse or rabbit serum proteins or serum proteins from bovine, horse, human, pig or rat, based on IEP. VisUCyteÖ HRP Polymer provides high sensitivity with low background in IHC. | |
| 6 months from date of receipt, 2 to 8 degreesC as supplied. |
| Immunohistochemistry | |
| HRP Polymer | |
| Supplied as a solution in PBS with preservative. | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3500 | |
| Mouse, Rabbit | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering