Learn More
STEMCELL Technologies Mouse Recombinant IGF-II
Insulin-like growth factor 2
Brand: STEMCELL Technologies 78221.1
Additional Details : Weight : 0.00110kg
Description
- Insulin-like growth factor 2 (IGF-II) is a polypeptide that belongs to the family of insulin-like growth factors that are similar in molecular structure to proinsulin
- IGF-II binding to IGF-I receptor activates a signaling cascade via the MAPK and PI3K pathways
- IGF-II affects growth, differentiation, and survival of cells in a variety of tissues during embryonic development
- It stimulates proliferation and migration of primary lymphatic endothelial cells, and induces lymphangiogenesis (Guvakova)
Specifications
Insulin-like growth factor type 2, Insulin-like growth factor II, Somatomedin A, T3M-11-derived growth factor | |
100 μg | |
Hematopoietic Stem and Progenitor Cells; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Prostate Cells | |
≥95% |