missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Mouse Recombinant IGF-II
Insulin-like growth factor 2
Specifications
Name | Insulin-like growth factor type 2, Insulin-like growth factor II, Somatomedin A, T3M-11-derived growth factor |
---|---|
Cell Type | Hematopoietic Stem and Progenitor Cells; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Prostate Cells |
Purity or Quality Grade | ≥95% |
Description
- Insulin-like growth factor 2 (IGF-II) is a polypeptide that belongs to the family of insulin-like growth factors that are similar in molecular structure to proinsulin
- IGF-II binding to IGF-I receptor activates a signaling cascade via the MAPK and PI3K pathways
- IGF-II affects growth, differentiation, and survival of cells in a variety of tissues during embryonic development
- It stimulates proliferation and migration of primary lymphatic endothelial cells, and induces lymphangiogenesis (Guvakova)
Specifications
Insulin-like growth factor type 2, Insulin-like growth factor II, Somatomedin A, T3M-11-derived growth factor | |
Hematopoietic Stem and Progenitor Cells; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Prostate Cells | |
≥95% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies Mouse Recombinant IGF-II