Learn More
STEMCELL Technologies Mouse Recombinant IGF-II
Insulin-like growth factor 2
Brand:  STEMCELL Technologies 78221.3
Additional Details : Weight : 0.00110kg
 View more versions of this product
Product Code. 17129891
  • POA / 1000µg
Estimated Shipment: 13-06-2024
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • Insulin-like growth factor 2 (IGF-II) is a polypeptide that belongs to the family of insulin-like growth factors that are similar in molecular structure to proinsulin
  • IGF-II binding to IGF-I receptor activates a signaling cascade via the MAPK and PI3K pathways
  • IGF-II affects growth, differentiation, and survival of cells in a variety of tissues during embryonic development
  • It stimulates proliferation and migration of primary lymphatic endothelial cells, and induces lymphangiogenesis (Guvakova)
Specifications
Specifications
Insulin-like growth factor type 2, Insulin-like growth factor II, Somatomedin A, T3M-11-derived growth factor
1000 μg
Hematopoietic Stem and Progenitor Cells; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Prostate Cells
≥95%