Learn More
STEMCELL Technologies Mouse Recombinant IL-4
Interleukin 4
2596.05 SEK - 42785.55 SEK
Specifications
Name | B cell stimulating factor, BCDF, BCGF, BSF-1, Interleukin-4 |
---|---|
Cell Type | B Cells; Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Lymphocytes; Monocytes; Myeloid Cells; Neurons; T Cells |
Purity or Quality Grade | ≥95% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17179431
![]() |
STEMCELL Technologies
78047 |
100 ÎĽg |
8557.65 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17189431
![]() |
STEMCELL Technologies
78047.1 |
20 ÎĽg |
2596.05 SEK
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17199431
![]() |
STEMCELL Technologies
78047.2 |
1000 ÎĽg |
42785.55 SEK
1000µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Interleukin 4 (IL-4) is a type I cytokine produced by Th2 T cells, NK T cells, γ/δ T cells, eosinophils, mast cells, and activated basophils
- IL-4 receptors are present on a number of cells, including hematopoietic, endothelial, epithelial, muscle, and brain tissues
- IL-4 receptor engagement initiates signaling through the JAK/STAT pathway, and leads to the activation of PI3K and Ras/MAPK pathways (Nelms et al.) IL-4 plays a key role in immunoglobulin class switching in B cells and in regulating the differentiation of naĂŻve T cells into T helper 2 (Th2) cells, while inhibiting Th1 differentiation
- IL-4 acts together with tumor necrosis factor to stimulate expression of adhesion molecules on vascular endothelial cells and to downregulate expression of E-selectin, thus recruiting T cells and eosinophils to the site of inflammation (Nelms et al.)
Specifications
B cell stimulating factor, BCDF, BCGF, BSF-1, Interleukin-4 | |
B Cells; Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Lymphocytes; Monocytes; Myeloid Cells; Neurons; T Cells | |
≥95% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies Mouse Recombinant IL-4