Learn More
STEMCELL Technologies Mouse Recombinant IL-4
Interleukin 4
Encompass_Preferred
2596.05 SEK - 42785.55 SEK
Specifications
Name B cell stimulating factor, BCDF, BCGF, BSF-1, Interleukin-4
Cell Type B Cells; Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Lymphocytes; Monocytes; Myeloid Cells; Neurons; T Cells
Purity or Quality Grade ≥95%
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17179431
Encompass_Preferred
View Documents
STEMCELL Technologies
78047
100 ÎĽg
8557.65 SEK
100µg
View alternative products
Save Up to 
»
17189431
Encompass_Preferred
View Documents
STEMCELL Technologies
78047.1
20 ÎĽg
2596.05 SEK
20µg
View alternative products
Save Up to 
»
17199431
Encompass_Preferred
View Documents
STEMCELL Technologies
78047.2
1000 ÎĽg
42785.55 SEK
1000µg
View alternative products
Save Up to 
»
Description
Description
  • Interleukin 4 (IL-4) is a type I cytokine produced by Th2 T cells, NK T cells, Îł/δ T cells, eosinophils, mast cells, and activated basophils
  • IL-4 receptors are present on a number of cells, including hematopoietic, endothelial, epithelial, muscle, and brain tissues
  • IL-4 receptor engagement initiates signaling through the JAK/STAT pathway, and leads to the activation of PI3K and Ras/MAPK pathways (Nelms et al.) IL-4 plays a key role in immunoglobulin class switching in B cells and in regulating the differentiation of naĂŻve T cells into T helper 2 (Th2) cells, while inhibiting Th1 differentiation
  • IL-4 acts together with tumor necrosis factor to stimulate expression of adhesion molecules on vascular endothelial cells and to downregulate expression of E-selectin, thus recruiting T cells and eosinophils to the site of inflammation (Nelms et al.)
Specifications
Specifications
B cell stimulating factor, BCDF, BCGF, BSF-1, Interleukin-4
B Cells; Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Lymphocytes; Monocytes; Myeloid Cells; Neurons; T Cells
≥95%
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies Mouse Recombinant IL-4