Learn More
R&D Systems™ Mouse TIM-1/KIM-1/HAVCR Quantikine ELISA Kit
Mouse TIM-1/KIM-1/HAVCR Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ MKM100
This item is not returnable.
View return policy
Description
The Mouse TIM-1/KIM-1/HAVCR Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse TIM-1/KIM-1/HAVCR in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), urine (10 ul).
Specifications
7.81 - 500pg/mL | |
HRP | |
Natural and Recombinant Mouse TIM-1. The antibody components of this kit were raised against the short splice form of Mouse TIM-1 (isoform 2) which has a 23 amino acid deletion following Pro182. This immunoassay detects both the short and long forms of Mouse TIM-1. | |
171283 | |
Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Urine (10uL) | |
CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1 TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 |
2.18pg/mL | |
Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine | |
ELISA | |
1 x 96-well Plate | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Content Correction
Product Title
R&D Systems™ Mouse TIM-1/KIM-1/HAVCR Quantikine ELISA Kit > One 96-well plate
Spot an opportunity for improvement?Share a Content Correction