Learn More
R&D Systems™ Mouse TIM-1/KIM-1/HAVCR Quantikine ELISA Kit
Mouse TIM-1/KIM-1/HAVCR Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand:  R&D Systems™ MKM100
Product Code. 18741924
  • 7735.00 SEK / 1 set
This item is not returnable. View return policy
Description
Description
The Mouse TIM-1/KIM-1/HAVCR Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse TIM-1/KIM-1/HAVCR in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), urine (10 ul).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
7.81 - 500pg/mL
HRP
Natural and Recombinant Mouse TIM-1. The antibody components of this kit were raised against the short splice form of Mouse TIM-1 (isoform 2) which has a 23 amino acid deletion following Pro182. This immunoassay detects both the short and long forms of Mouse TIM-1.
171283
Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Urine (10uL)
CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1 TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
2.18pg/mL
Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
ELISA
1 x 96-well Plate
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Mouse TIM-1/KIM-1/HAVCR Quantikine ELISA Kit > One 96-well plate