Learn More
MP Biomedicals™ Malate Dehydrogenase, From Porcine Heart (Mitochondrial)
Used for enzymatic determination of L-malate and gluamate oxaloacetate transaminase (GOT)
Brand: MP Biomedicals™ 0216004950
Additional Details : CAS Number : 9001-64-3 Weight : 0.01000kg
Description
Malate Dehydrogenase, [9001-64-3]
- CAS Number: 9001-64-3
- EC Number: 232-622-5
- Specific Activity: ∽1200u/mg protein
- Format: Powder
Specifications
≈1200 U/mg | |
Enzymatic determination of L-malate and Gluamate Oxaloacetate Transaminase | |
Porcine heart | |
Malate Dehydrogenase |
White | |
0°C | |
50 kU | |
Lyophilized Powder |