missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Malate Dehydrogenase, From Porcine Heart (Mitochondrial)
Used for enzymatic determination of L-malate and gluamate oxaloacetate transaminase (GOT)
1902.70 SEK - 6317.60 SEK
Specifications
Concentration | ≈1200 U/mg |
---|---|
Color | White |
For Use With (Application) | Enzymatic determination of L-malate and Gluamate Oxaloacetate Transaminase |
Content And Storage | 0°C |
Source | Porcine heart |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11440222
|
MP Biomedicals™
0216004910 |
10 kU |
1902.70 SEK
10ku |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11450222
|
MP Biomedicals™
0216004950 |
50 kU |
6317.60 SEK
50ku |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Malate Dehydrogenase, [9001-64-3]
- CAS Number: 9001-64-3
- EC Number: 232-622-5
- Specific Activity: ∽1200u/mg protein
- Format: Powder
Specifications
≈1200 U/mg | |
Enzymatic determination of L-malate and Gluamate Oxaloacetate Transaminase | |
Porcine heart | |
Lyophilized Powder |
White | |
0°C | |
Malate Dehydrogenase |