Learn More
MP Biomedicals™ Minimal Essential Medium Eagle (Modified) with Earle's Salts, without Sodium Bicarbonate and L-Glutamine
Product Code. 11440325 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 x 1 L
Unit Size:
10 L
Product Code. Quantity Unit Size
11440325 10 x 1 L 10 L
1 options
Product Code. 11440325 Supplier MP Biomedicals™ Supplier No. 091110020
  • ONLINE EXCLUSIVE PRICE
    577.00 SEK
    549.16 SEK / 10 L
    Save 27.84 SEK
    4% Av
 Restricted Item
I lager
Autoclavable Powder, MEM Modified
MEM, originally prepared by Harry Eagle, is one of the most popular cell culture media. Can be used to support the growth of cells in monolayers, in suspension and wide variety of other cell types with proper supplementation.
Specifications
Product Type MEM Eagle
Without Additives No Glutamine, No Sodium Bicarbonate
Quantity 10 x 1 L
Content And Storage 2 to 8°C
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.