missing translation for 'onlineSavingsMsg'
Learn More

MPP6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11575-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16888743

  • 3665.00 SEK / 150µL
Estimated Shipment: 30-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Members of the peripheral membrane-associated guanylate kinase (MAGUK) family function in tumor suppression and receptor clustering by forming multiprotein complexes containing distinct sets of transmembrane, cytoskeletal, and cytoplasmic signaling proteins. All MAGUKs contain a PDZ-SH3-GUK core and are divided into 4 subfamilies, DLG-like, ZO1-like, p55-like, and LIN2-like, based on their size and the presence of additional domains. MPP6 is a member of the p55-like MAGUK subfamily.
Specifications

Specifications

MPP6
Polyclonal
Unconjugated
MPP6
MAGUK p55 subfamily member 6, MPP6, p55T, PALS2, VAM 1, VAM1, Veli associated MAGUK 1
Rabbit
Antigen Affinity Chromatography
RUO
362359, 51678, 56524
-20°C
Liquid
Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9JLB0, Q9NZW5
Mpp6
MPP6 Fusion Protein Ag2158
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers