missing translation for 'onlineSavingsMsg'
Learn More

MRE11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen MRE11
Concentration 0.13 mg/mL
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16897323
View Documents
Proteintech
10744-1-AP-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16887323
View Documents
Proteintech
10744-1-AP-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Mre11 is an 80 kDa protein that acts as a Mn+2 dependent double stranded DNA 3′ to 5′ exonuclease and a single stranded DNA endonuclease. It is part of a larger complex with Rad50, p95 (Nibrin) and possibly other associated proteins involved in the nucleolytic end-processing of double strand breaks and telomere maintece.
Specifications

Specifications

MRE11
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Rat, Mouse, Human
P49959, Q61216
17535, 4361, 64046
MRE11 Fusion Protein Ag1105
Primary
-20°C
MRE11
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
ATLD, HNGS1, MRE11, MRE11 homolog 1, MRE11 homolog A, MRE11A, MRE11B
MRE11, Mre11a
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MRE11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.