missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17041-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that contains a high level of sequence similarity with ribosomal protein S11P family members. A pseudogene corresponding to this gene is found on chromosome 20. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
MRPS11 | |
Polyclonal | |
Unconjugated | |
MRPS11 | |
HCC 2, MRP S11, MRPS11, RPMS11, S11mt | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
499185, 64963, 67994 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P82912, Q9DCA2 | |
Mrps11 | |
MRPS11 Fusion Protein Ag10570 | |
150 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |