missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | MSH2 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16894724
|
Proteintech
15520-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16884724
|
Proteintech
15520-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Plays a role in neurogenesis and neuronal migration By similarity. Necessary for correct formation of mitotic spindles and chromosome separation during mitosis. Necessary for cytokinesis and cell proliferation.Specifications
MSH2 | |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
P43246, P43247, P54275 | |
17685, 4436, 81709 | |
MSH2 Fusion Protein Ag7835 | |
Primary | |
-20°C | |
MSH2 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
COCA1, FCC1, hMSH2, HNPCC, HNPCC1, LCFS2, MSH2, MutS protein homolog 2 | |
Msh2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MSH2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech