missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYBPC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19977-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The antibody detects the full length form and the degradation bands around 28-54 kDa in SDS-PAGE.
MYBPC3 encodes the cardiac isoform of myosin-binding protein C. Myosin-binding protein C is a myosin-associated protein found in the cross-bridge-bearing zone (C region) of A bands in striated muscle. MYBPC3, the cardiac isoform, is expressed exclussively in heart muscle. Regulatory phosphorylation of the cardiac isoform in vivo by cAMP-dependent protein kinase (PKA) upon adrenergic stimulation may be linked to modulation of cardiac contraction. Mutations in MYBPC3 are one cause of familial hypertrophic cardiomyopathy.Specifications
MYBPC3 | |
Polyclonal | |
Unconjugated | |
MYBPC3 | |
Cardiac MyBP C, CMH4, FHC, MYBP C, MYBPC3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17868, 295929, 4607 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O70468, P56741, Q14896 | |
Mybpc3 | |
Peptide | |
150 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |