missing translation for 'onlineSavingsMsg'
Learn More

MYCBP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12022-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16879483

  • 1355.00 SEK / 20µL
Estimated Shipment: 31-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This is a rabbit polyclonal antibody raised against full-length MYCBP of human origin.

The MYCBP gene encodes a protein that binds to the N-terminal region of MYC and stimulates the activation of E box-dependent transcription by MYC.
Specifications

Specifications

MYCBP
Polyclonal
Unconjugated
MYCBP
AMY 1, AMY1, Associate of Myc 1, c myc binding protein, MYCBP
Rabbit
Antigen Affinity Chromatography
RUO
100361133, 26292, 56309
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q99417, Q9EQS3
Mycbp
MYCBP Fusion Protein Ag2651
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers