missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH9 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | MYH9 |
---|---|
Concentration | 0.19 mg/mL |
Applications | Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887993
|
Proteintech
11128-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16877993
|
Proteintech
11128-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMP21 is a ubiquitously expressed protein that is involved in vesicular targeting and protein transport. More recent experiments have shown that it is also a component in the presenilin complex and modulates the -gamma-secretase but not the epsilon-secretase cleavage activity of the amyloid precursor protein. The presenilin complex is composed of the proteins APH1, nicastrin, and PEN2 in addition to presenilin-1. Together, these proteins cleave the amyloid precursor protein at what is known as the -gamma- and epsilon-sites and can lead to the accumulation of the Abeta cleavage product that is associated with Alzheimer′s disease. Co-immunoprecipitation experiments using antibodies against these proteins also yielded TMP21 indicating that TMP21 may play a role in the regulation of this complex. Suppression of TMP21 expression by siRNA in transfected cells caused increased -gamma-secretase activity but not epsilon-secretase activity, and increased Abeta production, demonstrating that TMP21 can modulate -gamma-secretase activity.Specifications
MYH9 | |
Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Zebrafish, Mouse | |
P35579, Q62812, Q8VDD5 | |
100911597, 17886, 333938, 4627 | |
MYH9 Fusion Protein Ag1612 | |
Primary | |
-20°C | |
MYH9 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
MYH9, Myosin 9, Myosin heavy chain 9, myosin IIa, Nonmuscle myosin IIa | |
Myh9, myh9a | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MYH9 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech