Learn More
MYO1D Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14028-1-AP-150UL
This item is not returnable.
View return policy
Specifications
MYO1D | |
Polyclonal | |
Unconjugated | |
MYO1D | |
KIAA0727, MYO1D, myosin ID, myr4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
338367, 4642 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O94832, Q5SYD0 | |
Myo1d | |
MYO1D Fusion Protein Ag5176 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Product Title
MYO1D Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction