Learn More
Myoglobin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16048-1-AP-150UL
 View more versions of this product
Product Code. 16885514
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
This antibody well recognized endogenous myoglobin in muscle lysates.
Myoglobin is a cytoplasmic, single chain polypeptide of 153 amino acids and contains a single heme group. Myoglobin is found in skeletal and carciac muscle but not in smooth muscle and functions as an oxygen transporting pigment. Upon damage to the muscle cell due to infarction of a coronary artery, neurological trauma, infection of tumor processes, myoglobin escapes to the environment and can be found in plasma using sensitive assays.
Specifications
Specifications
Myoglobin
Polyclonal
Unconjugated
MB
MB, MYO, myoglobin
Rabbit
Antigen Affinity Chromatography
RUO
17189, 4151, 59108
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P02144, P04247, Q9QZ76
Mb
Myoglobin Fusion Protein Ag9000
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Myoglobin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated