missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myoglobin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16048-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody well recognized endogenous myoglobin in muscle lysates.
Myoglobin is a cytoplasmic, single chain polypeptide of 153 amino acids and contains a single heme group. Myoglobin is found in skeletal and carciac muscle but not in smooth muscle and functions as an oxygen transporting pigment. Upon damage to the muscle cell due to infarction of a coronary artery, neurological trauma, infection of tumor processes, myoglobin escapes to the environment and can be found in plasma using sensitive assays.Specifications
Myoglobin | |
Polyclonal | |
Unconjugated | |
MB | |
MB, MYO, myoglobin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17189, 4151, 59108 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02144, P04247, Q9QZ76 | |
Mb | |
Myoglobin Fusion Protein Ag9000 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Myoglobin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction