Learn More
MYOM3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17692-1-AP-150UL
 View more versions of this product
Product Code. 16887624
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
This antibody recognizes the intact MYOM3 protein (160-160 kDa) as well as MYOM3 fragments (100 kDa and 130 kDa).
MYOM3 (myomesin 3) is a structural component of the M-band in striated muscle and is involved in sarcomere stability and resistance during intense or sustained stretching. MYOM3 can be detected mainly in intermediate speed fibers of skeletal muscle. Recently high level of MYOM3 fragments were detected in sera from patients with muscular dystrophy, including Duchenne muscular dystrophy (DMD). MYOM3 fragments may be used as serum biomarker for DMD and other neuromuscular disorders.
Specifications
Specifications
MYOM3
Polyclonal
Unconjugated
MYOM3
MYOM3, Myomesin 3, Myomesin family member 3, myomesin family, member 3, RP11 293P20.1
Rabbit
Antigen Affinity Chromatography
RUO
127294, 242702
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A2ABU4, Q5VTT5
Myom3
MYOM3 Fusion Protein Ag11849
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MYOM3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated