Läs mer
N-(2-Pyridyl)oxamic Acid, TRC
Produktkod. 30137961
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137961 1 g
17900335 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137961 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP9957001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13120-39-3
Molekylformel C7 H6 N2 O3
Molekylvikt (g/mol) 166.13
Synonym N-(2-Pyridinyl)oxamic Acid
IUPAC-namn 2-oxo-2-(pyridin-2-ylamino)acetic acid
LEDER OC(=O)C(=O)Nc1ccccn1
Specifikationer
Kemiskt namn eller material N-(2-Pyridinyl)oxamic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C7H6N2O3/c10-6(7(11)12)9-5-3-1-2-4-8-5/h1-4H,(H,11,12)(H,8,9,10)
Formel vikt 166.0378
Procent renhet >95%