CAS RN 13120-39-3
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
N-(2-Pyridinyl)oxamic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
N-(2-Pyridyl)oxamic Acid, TRC
CAS: 13120-39-3 Molekylformel: C7 H6 N2 O3 Molekylvikt (g/mol): 166.13 Synonym: N-(2-Pyridinyl)oxamic Acid IUPAC-namn: 2-oxo-2-(pyridin-2-ylamino)acetic acid LEDER: OC(=O)C(=O)Nc1ccccn1