Läs mer
N-[(4-Hydroxy-1-methyl-7-phenoxy-3-isoquinolinyl)carbonyl]glycine, TRC
Produktkod. 30137404
Change view
Kvantitet:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137404 250 mg
30007136 10 mg
30005512 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137404 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH948180250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 808118-40-3
Molekylformel C19 H16 N2 O5
Molekylvikt (g/mol) 352.34
Synonym Glycine, N-[(4-hydroxy-1-methyl-7-phenoxy-3-isoquinolinyl)carbonyl]-, N-[(4-Hydroxy-1-methyl-7-phenoxy-3-isoquinolinyl)carbonyl]glycine, (4-Hydroxy-1-methyl-7-phenoxyisoquinoline-3-carbonyl)glycine, ASP 1517, FG 4592, Roxadustat, [[(4-Hydroxy-1-methyl-7-phenoxyisoquinolin-3-yl)carbonyl]amino]acetic acid
IUPAC-namn 2-[(4-hydroxy-1-methyl-7-phenoxyisoquinoline-3-carbonyl)amino]acetic acid
LEDER Cc1nc(C(=O)NCC(=O)O)c(O)c2ccc(Oc3ccccc3)cc12
Specifikationer
Kemiskt namn eller material FG-4592
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C19H16N2O5/c1-11-15-9-13(26-12-5-3-2-4-6-12)7-8-14(15)18(24)17(21-11)19(25)20-10-16(22)23/h2-9,24H,10H2,1H3,(H,20,25)(H,22,23)
Kvantitet 250 mg
Formel vikt 352.1059
Procent renhet >95%