Läs mer
N-Methyl-p-tyramine, TRC
Produktkod. 17975065
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17975065 50 mg
17983845 10 mg
17940545 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17975065 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33235550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 370-98-9
Molekylformel C9 H13 N O
Molekylvikt (g/mol) 151.21
Synonym 4-Hydroxy-N-methylphenethylamine, Methyl-4-tyramine, p-(2-(Methylamino)ethyl)phenol
IUPAC-namn 4-[2-(methylamino)ethyl]phenol
LEDER CNCCc1ccc(O)cc1
Specifikationer
Kemiskt namn eller material N-Methyltyramine
Kvantitet 50 mg
InChI formel InChI=1S/C9H13NO/c1-10-7-6-8-2-4-9(11)5-3-8/h2-5,10-11H,6-7H2,1H3
Formel vikt 151.0997